missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Prodynorphin/PDYN Polyclonal specifically detects Prodynorphin/PDYN in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Classification | Polyclonal |
| Dilution | Western Blot 1.0 ug/ml |
| Molecular Weight of Antigen | 28 kDa |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?