Learn More
NLRP3/NALP3 Antibody (CL0210) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52879AF488
Product Code. 30291428
  • 737.00€ / 0.10mL
Description
NLRP3/NALP3 Monoclonal antibody specifically detects NLRP3/NALP3 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NLRP3/NALP3
Monoclonal
Alexa Fluor 488
AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1
This antibody was developed using a recombinant protein derived from Q96P20, with the exact immunogen sequence remaining proprietary.
0.1 mL
Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction
114548
Store at 4⪚C in the dark.
IgG1
Western Blot, Immunocytochemistry/Immunofluorescence
CL0210
50mM Sodium Borate
Mouse
Protein A purified