missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP2/NALP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58146-25ul
This item is not returnable.
View return policy
Description
NLRP2/NALP2 Polyclonal specifically detects NLRP2/NALP2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| NLRP2/NALP2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| CLR19.9, FLJ20510, NACHT, leucine rich repeat and PYD containing 2, NACHT, LRR and PYD domains-containing protein 2, NALP2NACHT, LRR and PYD containing protein 2, NBS1PYRIN-containing APAF1-like protein 2, NLR family, pyrin domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 2, PAN1Nucleotide-binding site protein 1, PYPAF2PYRIN domain and NACHT domain-containing protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| NLRP2 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VDVVPSSFCVKHCRNLQKMSLQVIKENLPENVTASESDAEVERSQDDQHMLPFWTDLCSIFGSNKDLMGLAINDSFLSASLVRILCEQ | |
| 25 μL | |
| Apoptosis | |
| 55655 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction