missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NLE1 plays a role in regulating Notch activity, in regulating the expression of CDKN1A and several members of the Wnt pathway, probably via its effects on Notch activity. NLE1 is required during embryogenesis for inner mass cell survival.
Specifications
Specifications
| Antigen | NLE1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.32 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | NLE1 |
| Gene Accession No. | Q8VEJ4, Q9NVX2 |
| Gene Alias | Nle, NLE1, Notchless protein homolog 1 |
| Gene Symbols | NLE1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?