Learn More
Tocris Bioscience™ NKY 80
GREENER_CHOICE
Product Code. 30002038
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 30002038
Brand: Tocris Bioscience™ 5071/50
Adenylyl cyclase inhibitor
Inhibitor of adenylyl cyclase (AC). Exhibits greater affinity for AC5 over AC3 and AC2 (IC50 values are 8.3 μM, 132 μM and 1.7 mM respectively).
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 299442-43-6
Molecular Formula C12H11N3O2
Specifications
Quantity 50 mg
Formula Weight 229.23
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 229.24
Chemical Name or Material NKY 80