CAS RN 299442-43-6
CAS RN 299442-43-6
IUPAC Name:
2-amino-7-(furan-2-yl)-5,6,7,8-tetrahydroquinazolin-5-one
Synonyms:
2-amino-7-(furan-2-yl)-7,8-dihydro-6H-quinazolin-5-one
Molecular Weight (g/mol):
229.24
Molecular Formula:
C12H11N3O2
InChi Key:
SOJUSNIBPPMLCC-UHFFFAOYNA-N
SMILES:
NC1=NC=C2C(=O)CC(CC2=N1)C1=CC=CO1