missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NKY 80

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Inhibitor of adenylyl cyclase (AC). Exhibits greater affinity for AC5 over AC3 and AC2 (IC50 values are 8.3 μM, 132 μM and 1.7 mM respectively).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 229.23 |
| Physical Form | Solid |
| Color | White |
| Purity | >98% |
| Molecular Weight (g/mol) | 229.24 |
| Chemical Name or Material | NKY 80 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction