missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX6.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97468-100ul
This item is not returnable.
View return policy
Description
NKX6.1 Polyclonal antibody specifically detects NKX6.1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| NKX6.1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Homeobox protein NK-6 homolog A, NK homeo box, family 6, member A, NK homeobox (Drosophila), family 6, A, NK homeobox, family 6, A, NK6 homeobox 1, NK6 transcription factor homolog A, NK6 transcription factor related, locus 1, NK6 transcription factor related, locus 1 (Drosophila), NKX6.1, NKX6Ahomeobox protein Nkx-6.1 | |
| Produced in rabbits immunized with E. coli-derived Human NKX6.1 fragment. | |
| 100 μg | |
| Pancreatic Stem Cell Markers, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
| 4825 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction