missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nkx2.2 Monoclonal Antibody (883411)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561919

Product Code. 30534582

  • 482.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nkx2.2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative
O95096, P42586
NKX2-2
E. coli-derived recombinant human NKX2.2, Met1-Lys128.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2b
Western Blot, Immunocytochemistry
883411
Unconjugated
NKX2-2
Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman
Mouse
Protein A/G
RUO
18088, 366214, 4821
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.