Learn More
NKp46/NCR1 Antibody (195314), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1850K
579.70 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18595299
  • 724.00€ / 0.10mL
Description
NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Agonist Activity, Immunocytochemistry, CyTOF-reported
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKp46/NCR1
Monoclonal
DyLight 488
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Agonist Activity, Immunocytochemistry, CyTOF
195314
Western Blot, Flow Cytometry, Immunohistochemistry, Agonist Activity, Immunocytochemistry, CyTOF-reported
CD335, CD335 antigen, hNKp46, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Mouse T cell hybridoma transfected with human NKp46/NCR1
0.1 mL
Adaptive Immunity, Immunology, Signal Transduction
9437
Store at 4°C in the dark.
IgG2b