Learn More
Invitrogen™ NKp30 Recombinant Rabbit Monoclonal Antibody (328)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560275
Product Code. 30536125
  • 429.00€ / 100µL
Description
NKp30 Recombinant Monoclonal Antibody for ELISA
The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells. The encoded protein interacts with CD3-zeta (CD247), a T-cell receptor. A single nucleotide polymorphism in the 5' untranslated region of this gene has been associated with mild malaria suceptibility. Three transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, May 2010].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKp30
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
O14931
Ncr3
Recombinant Human NKp30/NCR3 protein (Met1-Gly135).
100 μL
Primary
Human
Antibody
IgG
ELISA
328
Unconjugated
Ncr3
1C7; Activating natural killer receptor p30; activating NK-A1 receptor; CD337; LY117; lymphocyte antigen 117; MALS; Natural cytotoxicity triggering receptor 3; natural killer cell p30-related protein; natural killer cell receptor; Ncr3; NK receptor 1c7; Nkp30; NK-p30; NKp30S; NKp30v1; NKp30v1ED
Rabbit
Protein A
RUO
259197
-20°C, Avoid Freeze/Thaw Cycles