missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKG2C/CD159c/KLRC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 589.00€
Specifications
| Antigen | NKG2C/CD159c/KLRC2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18282935
|
Novus Biologicals
NBP2-58380 |
100 μL |
589.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633688
|
Novus Biologicals
NBP2-58380-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKG2C/CD159c/KLRC2 Polyclonal specifically detects NKG2C/CD159c/KLRC2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| NKG2C/CD159c/KLRC2 | |
| Polyclonal | |
| Rabbit | |
| Immunology | |
| CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor | |
| KLRC2 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 3822 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:CAVLQVNRLKSAQCGSSMIYHCKH | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title