missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK3R/TACR3/Neurokinin B Receptor Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
222.00€ - 507.00€
Specifications
| Antigen | NK3R/TACR3/Neurokinin B Receptor |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690141
|
Novus Biologicals
NBP2-93024-0.02ml |
0.02 mL |
222.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677521
|
Novus Biologicals
NBP2-93024-0.1ml |
0.1 mL |
507.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NK3R/TACR3/Neurokinin B Receptor Polyclonal antibody specifically detects NK3R/TACR3/Neurokinin B Receptor in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| NK3R/TACR3/Neurokinin B Receptor | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS (pH 7.3), 50% glycerol | |
| 6870 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Neurokinin B receptor, neurokinin beta receptor, neuromedin-K receptor, NK-3 receptor, NK-3R, NK3RMGC148061, NKR, TAC3R, TAC3RL, tachykinin receptor 3MGC148060 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 356-465 of human TACR3 (NP_001050.1). YCCLNKRFRAGFKRAFRWCPFIKVSSYDELELKTTRFHPNRQSSMYTVTRMESMTVVFDPNDADTTRSSRKKRATPRDPSFNGCSRRNSKSASATSSFISSPYTSVDEYS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title