missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrotyrosine Antibody (39B6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
197.00€
Specifications
| Antigen | Nitrotyrosine |
|---|---|
| Clone | 39B6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:4000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Microarray |
| Applications | Western Blot |
Description
Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Rat, Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.Specifications
| Nitrotyrosine | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat | |
| Hybridoma line 39B6 | |
| Primary | |
| Store at -20°C. |
| 39B6 | |
| Western Blot 1:4000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Microarray | |
| Monoclonal | |
| Purified | |
| RUO | |
| Nitrotyrosine | |
| IgG2a | |
| Protein G purified | |
| Recognizes 3-nitrotyrosine moieties. No detectable cross-reactivity with non-nitrated tyrosine. Not species specific. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title