Learn More
Nitrosamines Mixture 1013 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 4-Nitrosomorpholine∣N-Nitroso-diethylamine∣N-Nitrosodimethylamine∣N-Nitroso-di-n-butylamine∣N-Nitroso-di-n-propylamine∣N-Nitroso-diphenylamine∣N-Nitrosomethylethylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine |
| CAS | 59-89-2, 55-18-5, 62-75-9, 924-16-3, 621-64-7, 86-30-6 |
| Analyte or Component Names | 4-Nitrosomorpholine∣N-Nitroso-diethylamine∣N-Nitrosodimethylamine∣N-Nitroso-di-n-butylamine∣N-Nitroso-di-n-propylamine∣N-Nitroso-diphenylamine∣N-Nitrosomethylethylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine |
| Shipping Condition | Ice pack (-20°C) |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 4-Nitrosomorpholine 2000 μg/mL∣N-Nitroso-diethylamine 2000 μg/mL∣N-Nitrosodimethylamine 2000 μg/mL∣N-Nitroso-di-n-butylamine 2000 μg/mL∣N-Nitroso-di-n-propylamine 2000 μg/mL∣N-Nitroso-diphenylamine 2000 μg/mL∣N-Nitrosomethylethylamine 2000 μg/mL∣N-Nitrosopiperidine 2000 μg/mL∣N-Nitrosopyrrolidine 2000 μg/mL |