missing translation for 'onlineSavingsMsg'
Learn More

Nital Etchant, 2% (v/v) in Alcohol, Ricca Chemical

Product Code. 31940509
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940509 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 31940509 Supplier Ricca Chemical Company Supplier No. R53143001C
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Chemical Name or Material Nital Etchant, 2% (v/v) in Alcohol
Color Colorless
CAS 64-17-5, 67-56-1, 7697-37-2, 7732-18-5
CAS Min % 91.68, 4.58, 2.62, 1.12
CAS Max % 91.68, 4.58, 2.62, 1.12
Density 0.803
Solubility Information Miscible in water
Percent Purity 2%
Grade Laboratory
Odor Alcohol
Packaging Amber Glass Bottle
pH Acidic
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.