missing translation for 'onlineSavingsMsg'
Learn More

Nital Etchant, 10% (v/v) Nitric Acid in Reagent Alcohol, Ricca Chemical

Product Code. 31940520
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940520 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 31940520 Supplier Ricca Chemical Company Supplier No. R53230004A
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Chemical Name or Material Nital Etchant
Color Colorless to Yellow
CAS 64-17-5, 7697-37-2, 67-63-0, 67-63-0
CAS Min % 73.0, 16.92, 3.87
CAS Max % 76.0, 17.61, 4.03
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
pH <1
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 10%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.