Learn More
Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets
Description
pH measurements are only as accurate as the buffer solutions used for calibration purposes. Maximum precision is guaranteed with fully traceable NIST/DIN buffers. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
- NIST/DIN pH buffers are specified and guaranteed to three decimal places, offering top precision
- Individual sachets guarantee fresh solution for every calibration, as well as maximum ease of use
- A test certificate and detailed label with a temperature table comes with every box
- All SDS and labels contain information according to GHS (Globally Harmonized System)
Specifications
Specifications
| Description | NIST/DIN Buffer pH 9.180 sachets 30x20mL |
| For Use With (Equipment) | pH Electrode |
| For Use With (Application) | pH measurement, electrode calibration |
| NIST Traceable | Yes |
| Packaging | Sachet |
| pH Range | 9.180 (at 25°C) |
| Quantity | 30 x 20 mL |