missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35303-20ul
This item is not returnable.
View return policy
Description
NIR2 Polyclonal antibody specifically detects NIR2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| NIR2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DRES9NIR-2, Drosophila retinal degeneration B homolog, FLJ44997, membrane-associated phosphatidylinositol transfer protein 1, NIR2PITPNM, phosphatidylinositol transfer protein, membrane-associated 1Pyk2 N-terminal domain-interacting receptor 2, PITPnm 1, Rd9, RDGB, RDGB1, RDGBA, RDGBA1, retinal degeneration B alpha 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 110-190 of human NIR2 (NP_004901.2).,, Sequence:, EIETYYLPDGGQQPNVFNLSGAERRQRILDTIDIVRDAVAPGEYKAEEDPRLYHSVKTGRGPLSDDWARTAAQTGPLMCAY | |
| 20 μL | |
| Vision | |
| 9600 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction