Learn More
Niobium Standard: Nb @ 1000 μg/mL in 2% HF, VHG™
Product Code. 18081940
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18081940 100 mL 100mL
18061850 250 mL 250mL
18052840 50 mL 50mL
18050950 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 18081940 Supplier VHG Labs Supplier No. VHGPNBF100
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Niobium Standard: Nb @ 1000 μg/mL in 2% HF
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Niobium
CAS 3-1-7660
Methods ICP/ICP-MS
Analyte or Component Names Niobium
Shipping Condition Room Temperature
Molecular Formula Nb
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Nb
SMILES [Nb]
IUPAC Name Niobium
Solvent or Matrix Dilute HF
Molecular Weight (g/mol) 92
Quantity 100 mL
Formula Weight 92.9064 g/mol