missing translation for 'onlineSavingsMsg'
Learn More

Nifursol (~90%), TRC

Product Code. 17922355
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922355 10 mg 10mg
30132750 5 mg 5mg
30137888 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17922355 Supplier Toronto Research Chemicals Supplier No. TRCN45870010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16915-70-1
Formula Weight 365.0244
InChI Formula InChI=1S/C12H7N5O9/c18-11-8(3-6(15(20)21)4-9(11)16(22)23)12(19)14-13-5-7-1-2-10(26-7)17(24)25/h1-5,18H,(H,14,19)/b13-5+
IUPAC Name 2-hydroxy-3,5-dinitro-N-[(E)-(5-nitrofuran-2-yl)methylideneamino]benzamide
Molecular Formula C12 H7 N5 O9
Molecular Weight (g/mol) 365.21
SMILES Oc1c(cc(cc1[N+](=O)[O-])[N+](=O)[O-])C(=O)N\N=C\c2oc(cc2)[N+](=O)[O-]
Synonym 2-Hydroxy-3,5-dinitrobenzoic acid 2-[(5-nitro-2-furanyl)methylene]hydrazide, 2-Hydroxy-3,5-dinitrobenzoic acid [(5-nitro-2-furanyl)methylene]hydrazide, 3,5-Dinitrosalicylic acid (5-nitrofurfurylidene)hydrazide, Histomon, Nifursol, Salfuride, Sulfuride
Chemical Name or Material Nifursol

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.