missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niemann-Pick C1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-148MFV450
This item is not returnable.
View return policy
Description
Niemann-Pick C1 Polyclonal antibody specifically detects Niemann-Pick C1 in Human, Mouse, Rat, Porcine, Chinese Hamster, Hamster, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunomicroscopy, KnockDown, KnockDown
Specifications
| Niemann-Pick C1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| mFluor Violet 450 SE | |
| FLJ98532, Niemann-Pick C1 protein, Niemann-Pick disease, type C1, NPC, NPC1 | |
| A synthetic peptide made to the C-terminal region of human Niemann-Pick C. [UniProt# O15118] | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| 4864 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction