missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 5/CHRNA5 Antibody (2D2D10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
230.00€ - 590.00€
Specifications
| Antigen | Nicotinic Acetylcholine R alpha 5/CHRNA5 |
|---|---|
| Clone | 2D2D10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18277984
|
Novus Biologicals
NBP2-61677 |
0.1 mL |
590.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699428
|
Novus Biologicals
NBP2-61677-0.025ml |
0.025 mL |
230.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nicotinic Acetylcholine R alpha 5/CHRNA5 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 5/CHRNA5 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| Nicotinic Acetylcholine R alpha 5/CHRNA5 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| 1138 | |
| Purified recombinant fragment of human CHRNA5 (AA: extra 23-254) expressed in E. Coli. | |
| Primary | |
| 53 kDa |
| 2D2D10 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Signal Transduction | |
| Cholinergic receptor, neuronal nicotinic, alpha polypeptide-5, cholinergic receptor, nicotinic, alpha 5, cholinergic receptor, nicotinic, alpha polypeptide 5, LNCR2, NACHRA5, neuronal acetylcholine receptor subunit alpha-5, neuronal nicotinic acetylcholine receptor, alpha5 subunit | |
| CHRNA5 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title