missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 4/CHRNA4 Antibody (7B4H1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61674
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine R alpha 4/CHRNA4 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 4/CHRNA4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Nicotinic Acetylcholine R alpha 4/CHRNA4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| BFNC, cholinergic receptor, nicotinic, alpha 4, cholinergic receptor, nicotinic, alpha polypeptide 4, EBN, EBN1, ENFL1, FLJ95812, NACHR, NACHRA4, NACRA4, neuronal acetylcholine receptor subunit alpha-4, neuronal nicotinic acetylcholine receptor alpha-4 subunit | |
| Mouse | |
| 70 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 7B4H1 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CHRNA4 | |
| Purified recombinant fragment of human CHRNA4 (AA: extra 29-242) expressed in E. Coli. | |
| Protein G purified | |
| Neuroscience | |
| 1137 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction