Learn More
Nicotinamide N-Methyltransferase/NNMT Antibody (OTI3D8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand:  Novus Biologicals NBP2-00537
Product Code. 18706863
  • 575.00€ / 0.10mL
Description
Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Flow Cytometry
Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150
NNMT
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
EC 2.1.1.1, nicotinamide N-methyltransferase
Affinity Purified
IgG2b