Learn More
Nicosulfuron metabolite AUSN 100 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer
Product Code. 30324866
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324866 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30324866 Supplier Dr. Ehrenstorfer Supplier No. DREA15515200WL100
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Des-4,6-dimethoxypyrimidine Formimidamide Nicosulfuron
CAS 2307738-55-0
Analyte or Component Names Des-4,6-dimethoxypyrimidine Formimidamide Nicosulfuron
Shipping Condition +20°C
Molecular Formula C10 H14 N6 O4 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Nicosulfuron Metabolite AUSN, 3-Pyridinecarboxamide, 2-[[[[(aminoiminomethyl)amino]carbonyl]amino]sulfonyl]-N,N-dimethyl-, 2-[[[[(Aminoiminomethyl)amino]carbonyl]amino]sulfonyl]-N,N-dimethyl-3-Pyridinecarboxamide
InChI Formula InChI=1S/C10H14N6O4S/c1-16(2)8(17)6-4-3-5-13-7(6)21(19,20)15-10(18)14-9(11)12/h3-5H,1-2H3,(H5,11,12,14,15,18)
SMILES CN(C)C(=O)c1cccnc1S(=O)(=O)NC(=O)NC(=N)N
IUPAC Name 2-(carbamimidoylcarbamoylsulfamoyl)-N,N-dimethylpyridine-3-carboxamide
Solvent or Matrix Acetonitrile/Water
Molecular Weight (g/mol) 314.321
Quantity 1 mL
Formula Weight 314.0797 g/mol