Learn More
Nicorandil, 98+%, Thermo Scientific™
A vasodilator that activates ATP-sensitive potassium channels
72.70€ - 227.00€
Chemical Identifiers
| CAS | 65141-46-0 |
|---|---|
| Molecular Formula | C8H9N3O4 |
| Molecular Weight (g/mol) | 211.18 |
| MDL Number | MFCD00186520 |
| InChI Key | LBHIOVVIQHSOQN-UHFFFAOYSA-N |
| Synonym | nicorandil, ikorel, 2-nicotinamidoethyl nitrate, adancor, dancor, nicorandilum, sigmart, 2-pyridine-3-carbonylamino ethyl nitrate, n-2-hydroxyethyl nicotinamide nitrate, nicorandilum inn-latin |
| PubChem CID | 47528 |
| ChEBI | CHEBI:31905 |
| IUPAC Name | 2-[(pyridin-3-yl)formamido]ethyl nitrate |
| SMILES | [O-][N+](=O)OCCNC(=O)C1=CC=CN=C1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15443579
|
Thermo Scientific Alfa Aesar
J60879.MD |
250mg |
72.70€
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15433579
|
Thermo Scientific Alfa Aesar
J60879.03 |
1g |
227.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 65141-46-0 | |
| 211.18 | |
| LBHIOVVIQHSOQN-UHFFFAOYSA-N | |
| 47528 | |
| 2-[(pyridin-3-yl)formamido]ethyl nitrate |
| C8H9N3O4 | |
| MFCD00186520 | |
| nicorandil, ikorel, 2-nicotinamidoethyl nitrate, adancor, dancor, nicorandilum, sigmart, 2-pyridine-3-carbonylamino ethyl nitrate, n-2-hydroxyethyl nicotinamide nitrate, nicorandilum inn-latin | |
| CHEBI:31905 | |
| [O-][N+](=O)OCCNC(=O)C1=CC=CN=C1 |
Specifications
| 65141-46-0 | |
| MFCD00186520 | |
| nicorandil, ikorel, 2-nicotinamidoethyl nitrate, adancor, dancor, nicorandilum, sigmart, 2-pyridine-3-carbonylamino ethyl nitrate, n-2-hydroxyethyl nicotinamide nitrate, nicorandilum inn-latin | |
| [O-][N+](=O)OCCNC(=O)C1=CC=CN=C1 | |
| 211.18 | |
| CHEBI:31905 | |
| ≥98% |
| C8H9N3O4 | |
| 14,6521 | |
| LBHIOVVIQHSOQN-UHFFFAOYSA-N | |
| 2-[(pyridin-3-yl)formamido]ethyl nitrate | |
| 47528 | |
| 211.17 | |
| Nicorandil |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
missing translation for 'einecsNumber' : 265-514-1
missing translation for 'rtecsNumber' : US4667600
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.