Learn More
Nickel Electro™ Nickel 201 Dish Lids
Product Code. 12747376 Shop All Nickel-Electro Products
Change view
Click to view available options
Diameter (Metric):
55 mm
60 mm
70 mm
75 mm
85 mm
90 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
12747376 60 mm Each
12787376 90 mm Each
12777376 85 mm Each
12737376 55 mm Each
12757376 70 mm Each
12767376 75 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12747376 Supplier Nickel Electro™ Supplier No. 543
  • ONLINE EXCLUSIVE PRICE
    22.15€
    19.95€ / Pack of 1
    Save 2.20€
    9% Off
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Nickel Electro™ Nickel 201 Dish Lids are made to resist corrosive enviroments, which make it useful for maintaining product purity.
  • 99.6% nickel purity
  • Crevice free
  • High thermal and electrical conductivities
  • Low gas content
  • Low vapor pressure
  • Ideal for lab crucibles that are capable of withstanding oxidizing furnace atmospheres up to 1100°C
Specifications
Material Nickel
Type Dish Lid
Diameter (Metric) 60 mm