Learn More
Nicastrin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14071-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes the largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene contains a carboxy terminal domain composed of heptapeptide repeats that are essential for polymerase activity. These repeats contain serine and threonine residues that are phosphorylated in actively transcribing RNA polymerase. In addition, this subunit, in combination with several other polymerase subunits, forms the DNA binding domain of the polymerase, a groove in which the DNA template is transcribed into RNA.Specifications
| Nicastrin | |
| Polyclonal | |
| Unconjugated | |
| NCSTN | |
| APH2, KIAA0253, NCSTN, nicastrin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23385, 289231, 59287 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P57716, Q8CGU6, Q92542 | |
| Ncstn | |
| Nicastrin Fusion Protein Ag5137 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |