Learn More
Nicardipine hydrochloride, 98%, Thermo Scientific™
L-type calcium channel blocker
71.70€ - 264.00€
Chemical Identifiers
| CAS | 54527-84-3 |
|---|---|
| Molecular Formula | C26H30ClN3O6 |
| Molecular Weight (g/mol) | 515.991 |
| MDL Number | MFCD00057327 |
| InChI Key | AIKVCUNQWYTVTO-UHFFFAOYSA-N |
| Synonym | nicardipine hydrochloride, cardene, nicardipine hcl, perdipine, nicodel, loxen, angioglebil, bionicard, nicardil, perdipina |
| PubChem CID | 41114 |
| IUPAC Name | 5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride |
| SMILES | CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15453879
|
Thermo Scientific Alfa Aesar
J61269.03 |
1g |
71.70€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15463879
|
Thermo Scientific Alfa Aesar
J61269.06 |
5g |
264.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 54527-84-3 | |
| 515.991 | |
| AIKVCUNQWYTVTO-UHFFFAOYSA-N | |
| 41114 | |
| CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl |
| C26H30ClN3O6 | |
| MFCD00057327 | |
| nicardipine hydrochloride, cardene, nicardipine hcl, perdipine, nicodel, loxen, angioglebil, bionicard, nicardil, perdipina | |
| 5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride |
Specifications
| 54527-84-3 | |
| MFCD00057327 | |
| 14,6495 | |
| AIKVCUNQWYTVTO-UHFFFAOYSA-N | |
| 5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride | |
| 41114 | |
| 98% |
| C26H30ClN3O6 | |
| UN2811 | |
| nicardipine hydrochloride, cardene, nicardipine hcl, perdipine, nicodel, loxen, angioglebil, bionicard, nicardil, perdipina | |
| CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl | |
| 515.991 | |
| 515.99 | |
| Nicardipine hydrochloride |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
H301+H311-H330
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 259-198-4
missing translation for 'rtecsNumber' : US7972100
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.