missing translation for 'onlineSavingsMsg'
Learn More

Nicardipine hydrochloride, 98%, Thermo Scientific™

L-type calcium channel blocker

Brand:  Thermo Scientific Alfa Aesar J61269.03

Quantity 1g

 View more versions of this product

Product Code. 15453879

  • 71.70€ / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

54527-84-3
515.991
AIKVCUNQWYTVTO-UHFFFAOYSA-N
41114
CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl
C26H30ClN3O6
MFCD00057327
nicardipine hydrochloride, cardene, nicardipine hcl, perdipine, nicodel, loxen, angioglebil, bionicard, nicardil, perdipina
5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride

Specifications

54527-84-3
MFCD00057327
UN2811
nicardipine hydrochloride, cardene, nicardipine hcl, perdipine, nicodel, loxen, angioglebil, bionicard, nicardil, perdipina
CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl
515.991
515.99
Nicardipine hydrochloride
C26H30ClN3O6
1g
14,6495
AIKVCUNQWYTVTO-UHFFFAOYSA-N
5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
41114
98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.