Learn More
Nicametate, TRC
Product Code. 30416243
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416243 100 mg 100mg
30413279 500 mg 500mg
30408865 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30416243 Supplier Toronto Research Chemicals Supplier No. TRCN900825100MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3099-52-3
Molecular Formula C12 H18 N2 O2
Molecular Weight (g/mol) 222.28
Synonym Nicotinic Acid 2-Diethylamino-Ethyl Ester, 3-Pyridinecarboxylic acid, 2-(diethylamino)ethyl ester, Nicotinic acid, 2-(diethylamino)ethyl ester (7CI,8CI), 2-(Diethylamino)ethyl 3-pyridinecarboxylate, 2-(Diethylamino)ethyl nicotinate, Nicametate, beta-Diethylaminoethyl nicotinate
IUPAC Name 2-(diethylamino)ethyl pyridine-3-carboxylate
SMILES CCN(CC)CCOC(=O)c1cccnc1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C12H18N2O2/c1-3-14(4-2)8-9-16-12(15)11-6-5-7-13-10-11/h5-7,10H,3-4,8-9H2,1-2H3
Formula Weight 222.1368
Percent Purity >95%
Chemical Name or Material Nicametate
Grade HPLC