CAS RN 3099-52-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nicametate, TRC
CAS: 3099-52-3 Molecular Formula: C12 H18 N2 O2 Molecular Weight (g/mol): 222.28 Synonym: Nicotinic Acid 2-Diethylamino-Ethyl Ester,3-Pyridinecarboxylic acid, 2-(diethylamino)ethyl ester,Nicotinic acid, 2-(diethylamino)ethyl ester (7CI,8CI),2-(Diethylamino)ethyl 3-pyridinecarboxylate,2-(Diethylamino)ethyl nicotinate,Nicametate,beta-Diethylaminoethyl nicotinate IUPAC Name: 2-(diethylamino)ethyl pyridine-3-carboxylate SMILES: CCN(CC)CCOC(=O)c1cccnc1