missing translation for 'onlineSavingsMsg'
Learn More

NHLRC2 Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

129.00€ - 349.00€

Specifications

Antigen NHLRC2
Concentration 700 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234488
View Documents
Proteintech
29416-1-AP-150UL
150 μL
ONLINE EXCLUSIVE PRICE
410.00€
349.00€ / 150µL
Save 61.00€
14% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30235428
View Documents
Proteintech
29416-1-AP-20UL
20 μL
ONLINE EXCLUSIVE PRICE
145.00€
129.00€ / 20µL
Save 16.00€
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The NHLRC2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NHLRC2. This antibody recognizes human, mouse antigen. The NHLRC2 antibody has been validated for the following applications: WB, IHC, ELISA analysis.
Specifications

Specifications

NHLRC2
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Rabbit
Human, Mouse
DKFZp779F115; FLJ20147; FLJ25621; FLJ33312; MGC45492
IgG
Antigen affinity purification
Polyclonal Antibody
700 μg/mL
Polyclonal
Liquid
RUO
374354
Recombinant Protein
Primary
Store at -20°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.