missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHERF-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35762-100ul
This item is not returnable.
View return policy
Description
NHERF-2 Polyclonal antibody specifically detects NHERF-2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| NHERF-2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| E3KARP, Na(+)/H(+) exchange regulatory cofactor NHE-RF2, NHE3RF2, NHERF2MGC104639, NHERF-2NHE3 kinase A regulatory protein E3KARP, OCTS2, SIP-1SRY-interacting protein 1, sodium/hydrogen exchanger, Sodium-hydrogen exchanger regulatory factor 2, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 2, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatoryfactor 2, solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2, Solute carrier family 9 isoform A3 regulatory factor 2, TKA-1SIP1, Tyrosine kinase activator protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 180-300 of human NHERF-2 (NP_001123484).,, Sequence:, DPGSPAARSGLRAQDRLIEVNGQNVEGLRHAEVVASIKAREDEARLLVVDPETDEHFKRLRVTPTEEHVEGPLPSPVTNGTSPAQLNGGSACSSRSDLPGSDKDTEDGSAWKQDPFQESGL | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9351 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction