Learn More
Invitrogen™ NHEDC2 Monoclonal Antibody (896151)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561950
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Electroneutral exchange of protons for Na(+) and Li(+) across the inner mitochondrial membrane. Contributes to the organellar volume homeostasis. Required for osteoclast differentiation and bone resorption activity.
Specifications
| NHEDC2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q86UD5 | |
| SLC9B2 | |
| NS0 mouse myeloma cell line transfected with human NHA2/SLC9B2, Met1-Val537. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 896151 | |
| Unconjugated | |
| SLC9B2 | |
| C80638; mitochondrial Na(+)/H(+) exchanger NHA2; mitochondrial sodium/hydrogen exchanger 9B2; mitochondrial sodium/hydrogen exchanger NHA2; Na(+)/H(+) exchanger NHA2; Na(+)/H(+) exchanger-like domain-containing protein 2; Na+/H+ exchanger domain containing 2; Nha2; nhaoc; nha-oc; NHE domain-containing protein 2; NHE10; Nhedc2; SLC9B2; Soditlum/hydrogen exchanger-like domain-containing protein 2; Sodium/hydrogen exchanger 9B2; Sodium/hydrogen exchanger-like domain-containing protein 2; solute carrier family 9 member B2; solute carrier family 9 subfamily B member 2; solute carrier family 9, subfamily B (cation proton antiporter 2), member 2; solute carrier family 9, subfamily B (NHA2, cation proton antiporter 2), member 2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 133308 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |