missing translation for 'onlineSavingsMsg'
Learn More

NHA2/SLC9B2/NHEDC2 Antibody (896151) [DyLight 405], Novus Biologicals™

Product Code. 18533888 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18533888 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18533888 Supplier Novus Biologicals Supplier No. FAB8404E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NHA2/SLC9B2/NHEDC2 Monoclonal antibody specifically detects NHA2/SLC9B2/NHEDC2 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHA2/SLC9B2/NHEDC2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 896151
Conjugate DyLight 405
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ23984, Mitochondrial Na(+)/H(+) exchanger NHA2, mitochondrial sodium/hydrogen exchanger NHA2, Na(+)/H(+) exchanger-like domain-containing protein 2, Na+/H+ exchanger domain containing 2, NHA2NHE10, NHE domain-containing protein 2, Sodium/hydrogen exchanger-like domain-containing protein 2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NHA2/SLC9B2, Met1-Val537, Accession # NP_849155
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133308
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.