missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NgR3/NgRH2/RTN4RL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
415.00€
Specifications
| Antigen | NgR3/NgRH2/RTN4RL1 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NgR3/NgRH2/RTN4RL1 Polyclonal specifically detects NgR3/NgRH2/RTN4RL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NgR3/NgRH2/RTN4RL1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DKFZp547J144, NgR3, NGRH2, NGRL2, nogo receptor-like 2, Nogo-66 receptor homolog 2, Nogo-66 receptor-related protein 3, reticulon 4 receptor-like 1, reticulon-4 receptor-like 1 | |
| RTN4RL1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q86UN2 | |
| 146760 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AVPCVSPGLRHGQDLKLLRAEDFRNCTGPASPHQIKSHTLTTTDRAARKEHHSPHGPTRSKGHPHGPRPGHRKPGKNCTNPRNRNQISKAGAGKQAPELPDYAPDYQHKFSFD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title