missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGDN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16524-1-AP-150UL
This item is not returnable.
View return policy
Description
Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.Specifications
| NGDN | |
| Polyclonal | |
| Unconjugated | |
| NGDN | |
| C14orf120, EIF4E binding protein, LCP5, lpd 2, Neuroguidin, NGD, NGDN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25983, 305887, 68966 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NEJ9, Q9DB96 | |
| Ngdn | |
| NGDN Fusion Protein Ag9763 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction