missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGDN Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00€ - 470.00€
Specifications
| Antigen | NGDN |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607090
|
Novus Biologicals
NBP2-94497-0.02ml |
0.02 mL |
188.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621401
|
Novus Biologicals
NBP2-94497-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NGDN Polyclonal antibody specifically detects NGDN in Human, Rat samples. It is validated for Western BlotSpecifications
| NGDN | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| C14orf120, DKFZp564O092, EIF4E-binding protein, eukaryotic initiation factor 4E and CPEB binding protein, LCP5, lpd-2, neuroguidin, neuroguidin, EIF4E binding protein, NGDchromosome 14 open reading frame 120 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 61-209 of human NGDN (NP_001036100.1). LMYLMDLTHLILDKASGGSLQGHDAVLRLVEIRTVLEKLRPLDQKLKYQIDKLIKTAVTGSLSENDPLRFKPHPSNMMSKLSSEDEEEDEAEDDQSEASGKKSVKGVSKKYVPPRLVPVHYDETEAEREKKRLERAKRRALSSSVIREL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 25983 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title