missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFU1 iron-sulfur cluster scaffold homolog Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95100-0.1ml
This item is not returnable.
View return policy
Description
NFU1 iron-sulfur cluster scaffold homolog Polyclonal antibody specifically detects NFU1 iron-sulfur cluster scaffold homolog in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NFU1 iron-sulfur cluster scaffold homolog | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| CGI-33, mitochondrial, Nfu, NFU1 iron-sulfur cluster scaffold homolog (S. cerevisiae) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human NFU1 (NP_001002755.1). MAATARRGWGAAAVAAGLRRRFCHMLKNPYTIKKQPLHQFVQRPLFPLPAAFYHPVRYMFIQTQDTPNPNSLKFIPGKPVLETRTMDFPTPAAAFRSPLARQLFRIEGVKSVFFGPDFITVTKENEELDWNLLKPDIYATIMDFFASGLPLVTEETPSGEAGSEEDDEVVAMIKELLDTRIRPTVQEDGGDVIYKGFEDGIVQLKLQGSCTSCPSSIITLKNGIQNMLQF | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 27247 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction