Learn More
NFRKB Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-93126-0.1ml
 View more versions of this product
Product Code. 18660141
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
NFRKB Polyclonal antibody specifically detects NFRKB in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NFRKB
Polyclonal
Western Blot 1:500-1:2000
DKFZp547B2013, DNA-binding protein R kappa-B, INO80 complex subunit G, INO80GDNA-binding protein R kappa B, nuclear factor related to kappaB binding protein, nuclear factor related to kappa-B-binding protein
Recombinant fusion protein containing a sequence corresponding to amino acids 1-340 of human NFRKB (NP_001137307.1). MDSLDHMLTDPLELGPCGDGHGTRIMEDCLLGGTRVSLPEDLLEDPEIFFDVVSLSTWQEVLSDSQREHLQQFLPQFPEDSAEQQNELILALFSGENFRFGNPLHIAQKLFRDGHFNPEVVKYRQLCFKSQYKRYLNSQQQYFHRLLKQILASRSDLLEMARRSGPALPFRQKRPSPSRTPEEREWRTQQRYLKVLREVKEECGDTALSSDEEDLSSWLPSSPARSPSPAVPLRVVPTLSTTDMKTADKVELGDSDLKIMLKKHHEKRKHQPDHPDLLTGDLTLNDIMTRVNAGRKGSLAALYDLAVLKKKVKEKEEKKKKKIKTIKSEAEDLAEPLSST
0.1 mL
Primary
Human
Purified
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
4798
Store at -20°C. Avoid freeze-thaw cycles.