missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB2/NFkB p100 [p Ser866] Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33274-20ul
This item is not returnable.
View return policy
Description
NFkB2/NFkB p100 Monoclonal antibody specifically detects NFkB2/NFkB p100 in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| NFkB2/NFkB p100 | |
| Monoclonal | |
| Western Blot 1:100 - 1:500, ELISA Recommended starting concentration is 1 μg/mL | |
| CVID10;H2TF1;LYT10;LYT-10;NF-kB2;Nuclear factor NF-kappa-B p100 subunit;p100;p49/p100;p52 | |
| A synthetic phosphorylated peptide around S866 of human NFkB2/NFkB p100 (NP_001070962.1).,, Sequence:, EDSAY | |
| 20 μL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4791 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction