Learn More
NFkB2/NFkB p100 Antibody (CL8266) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88914AF532
Product Code. 30307619
  • 737.00€ / 0.10mL
Description
NFkB2/NFkB p100 Monoclonal antibody specifically detects NFkB2/NFkB p100 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NFkB2/NFkB p100
Monoclonal
Alexa Fluor 532
CVID10;H2TF1;LYT10;LYT-10;NF-kB2;Nuclear factor NF-kappa-B p100 subunit;p100;p49/p100;p52
This antibody was developed using a synthetic peptide derived from Q00653, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Immunology, Signal Transduction
4791
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL8266
50mM Sodium Borate