missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFATC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35757-20ul
This item is not returnable.
View return policy
Description
NFATC4 Polyclonal antibody specifically detects NFATC4 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| NFATC4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| NF-AT3, NFAT3nuclear factor of activated T-cells, cytoplasmic 4, NFATc4, NF-ATc4, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4, T cell transcription factor NFAT3, T-cell transcription factor NFAT3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 560-710 of human NFATC4 (NP_004545.2).,, Sequence:, QGGGKVVSVQAASVPIECSQRSAQELPQVEAYSPSACSVRGGEELVLTGSNFLPDSKVVFIERGPDGKLQWEEEATVNRLQSNEVTLTLTVPEYSNKRVSRPVQVYFYVSNGRRKRSPTQSFRFLPVICKEEPLPDSSLRGFPSASATPFG | |
| 20 μL | |
| Transcription Factors and Regulators, Wnt Signaling Pathway | |
| 4776 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction