Learn More
NF-M Antibody (CL2678) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46616JF646
Product Code. 30296533
  • 737.00€ / 0.10mL
Description
NF-M Monoclonal antibody specifically detects NF-M in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NF-M
Monoclonal
Janelia Fluor 646
NEF3neurofilament medium polypeptide, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein
This antibody was developed using a recombinant protein derived from P07197, with the exact immunogen sequence remaining proprietary.
0.1 mL
Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
4741
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL2678
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat