missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ New Zealand Bovine Transferrin (Apo), Heat-treated
Shop All MP Biomedical Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Bovine Transferrin (APO) is an iron-depleted product supplied as a heat-treated, lyophilized powder. Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
- Frequently used as a nutrient in cell and microbial culture
- Used as a media supplement in biopharmaceutical production
- Useful transport factor in cell culture work to convey essential metabolites to the cells
- Important factor when culturing cells in a defined medium
Specifications
Specifications
| For Use With (Application) | Media Nutrient Supplement |
| Molecular Weight (g/mol) | 76,000 to 81,000 g/mol |
| Quantity | 100 mg |
| Regulatory Status | Research Use Only |
| Species | Bovine |
| Recombinant | Native |
| Form | Powder |
| Purity or Quality Grade | Cell Culture Grade |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction