Learn More
Nevirapine EP Impurity B, Mikromol™
Description
Nevirapine EP Impurity B
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one |
| CAS | 287980-84-1 |
| Analyte or Component Names | 4-Methyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H10 N4 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Nevirapine Anhydrous Imp. B (EP), Nevirapine USP Related Compound B, Nevirapine USP RC B, 4-Methyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one, 6H-Dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one, 5,11-dihydro-4-methyl-, BIRG 616 BS, Nevirapine Related Compound B, Nevirapine Impurity B |
| InChI Formula | InChI=1S/C12H10N4O/c1-7-4-6-14-11-9(7)15-12(17)8-3-2-5-13-10(8)16-11/h2-6H,1H3,(H,15,17)(H,13,14,16) |
| SMILES | Cc1ccnc2Nc3ncccc3C(=O)Nc12 |
| IUPAC Name | 4-methyl-5,11-dihydrodipyrido[2,3-b:3',2'-e][1,4]diazepin-6-one |
| Show More |