missing translation for 'onlineSavingsMsg'
Learn More

NeutraControl VEGF165 Mouse Monoclonal Antibody, Proteintech

Product Code. 30234280 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30234280 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30234280 Supplier Proteintech Supplier No. 695251IG100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The NeutraControl VEGF165 antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraControl VEGF165. This antibody recognizes human antigen. The NeutraControl VEGF165 antibody has been validated for the following applications: ELISA, Non-Neutralization analysis.

Specifications

Antigen VEGF165
Applications ELISA, Non-neutralization
Classification Monoclonal
Clone 7E9F4
Conjugate Unconjugated
Gene Symbols MGC70609; MVCD1; VEGF; VEGF-A; VPF
Host Species Mouse
Immunogen RecombinantProtein
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7422
Target Species Human
Content And Storage Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Product Type Neu Control
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.