missing translation for 'onlineSavingsMsg'
Learn More

Neuron-specific beta-III Tubulin Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18737549
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18737549 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18737549 Supplier R&D Systems Supplier No. MAB1195SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Multi-Species samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-III Tubulin
Applications Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone TuJ-1
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 1-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias Beta3-tubulin, beta-4, betaIII Tubulin, beta-Tubulin III, Class III beta-tubulin, TUBB3, TUBB4 CFEOM3A, Tubulin Beta 3, tubulin beta-3 chain, Tubulin beta-4, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
Gene Symbols TUBB3
Host Species Mouse
Immunogen Rat brain-derived microtubules
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10381
Test Specificity Detects mammalian and chicken neuron-specific beta-III tubulin but not other beta-tubulin isotypes in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.